Anti-Tissue Plasminogen Activator (Ab-1) (GMA-043) Mouse Monoclonal Antibody [clone: GMA-043]

Proveedor: Merck Millipore (Oncogene)
GF53-100
GF53-100EA 448 EUR
GF53-100
Anti-Tissue Plasminogen Activator (Ab-1) (GMA-043) Mouse Monoclonal Antibody [clone: GMA-043]
Anticuerpos
Type: Primary
Antigen: Tissue Plasminogen Activator (Ab-1) (GMA-043)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR