Single channel pipettes, mechanical, variable volume, PIPETMAN® L
Proveedor: GILSON
FA10008 FA10007 FA10006M FA10003P FA10005P FA10006P FA10003M FA10005M FA10004P FA10004M FA10001M FA10001P FA10002M FA10002P F167350 F167550 F167370 FA10001MC FA10001MCG FA10001PC FA10001PCG FA10002MC FA10002MCG FA10002PC FA10002PCG FA10003MC FA10003MCG FA10003PC FA10003PCG FA10004MC FA10004MCG FA10004PC FA10004PCG FA10005MC FA10005MCG FA10005PC FA10005PCG FA10006MC FA10006MCG FA10006PC FA10006PCG FA10007C FA10007CG FA10008C FA10008CG
613-5813EA 342 EUR
613-5813 613-5812 613-5805 613-5808 613-5810 613-5811 613-5802 613-5804 613-5809 613-5803 613-5800 613-5806 613-5801 613-5807 613-5890 613-5891 613-5892 GILSFA10001MC GILSFA10001MCG GILSFA10001PC GILSFA10001PCG GILSFA10002MC GILSFA10002MCG GILSFA10002PC GILSFA10002PCG GILSFA10003MC GILSFA10003MCG GILSFA10003PC GILSFA10003PCG GILSFA10004MC GILSFA10004MCG GILSFA10004PC GILSFA10004PCG GILSFA10005MC GILSFA10005MCG GILSFA10005PC GILSFA10005PCG GILSFA10006MC GILSFA10006MCG GILSFA10006PC GILSFA10006PCG GILSFA10007C GILSFA10007CG GILSFA10008C GILSFA10008CG
Single channel pipettes, mechanical, variable volume, PIPETMAN® L
Pipetas Micropipetas
Designed for performance and comfort, PIPETMAN® L comes in a comprehensive range of models, adapted to your needs in the lab. PIPETMAN L air-displacement pipettes cover a full volume range. Comfortable, precise, accurate, and reliable, use PIPETMAN L pipettes confidently in your applications.
  • Convenient use
  • Secure pipetting
  • GLP compliance
Lightweight and balanced, PIPETMAN L combines an ergonomic design with minimized pipetting forces, giving you the ability to pipette longer without fatigue. Right-handed or left-handed? No problem. Adjust theMore Product Information