Anti-Glucocorticoid Receptor (Ab-2) (BuGR2) Mouse Monoclonal Antibody [clone: BuGR2]

Proveedor: Merck Millipore (Oncogene)
GR32L-100
GR32L-100EA 589 EUR
GR32L-100
Anti-Glucocorticoid Receptor (Ab-2) (BuGR2) Mouse Monoclonal Antibody [clone: BuGR2]
Anticuerpos
Type: Primary
Antigen: Glucocorticoid Receptor (Ab-2) (BuGR2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

ESPECIFICACIONES

Simbolo antígeno GCR
Environmentally Preferable
Nombre del antígeno Glucocorticoid Receptor
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado Unconjugated
Clon BuGR2
Reactividad
Huésped Mouse
Isotipo
Western blot Yes
InmunoQuímica Yes
Precipitación inmunológica Yes
FluorescenciaInmunológica Yes
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR