Mass cell culture system, CELLdisc™
Proveedor: Greiner Bio-One
678104_4 678904_4 678108_3 678908_3 678116_2 678916_2 678140_1 678940_1
GREI678104_4EA 392 EUR
GREI678104_4 GREI678904_4 GREI678108_3 GREI678908_3 GREI678116_2 GREI678916_2 GREI678140_1 GREI678940_1
Mass cell culture system, CELLdisc™
Biorreactores
CELLdisc™ is a multilayer device covering a range of cell culture surfaces from 250 cm² up to 10000 cm². The innovative, ergonomic design provides a versatile system from research scale to industrial batches.
  • Non pyrogenic, non cytotoxic
  • Free from DNase, RNase and human DNA
  • Screw cap colour indicating surface treatment
  • Integrated gas support channel and vent port for pressure equilibration
  • Available as 4-, 8-, 16- and 40-layer version
A centrally located channel allows uniform distribution of gas throughout the device. A socle-rim guarantees that theMore Product Information