Biobanking tubes, Cryo.s™
Proveedor: Greiner Bio-One
978561_960 978570_960 978580_960 976561_960 976570_10 976567_960 977561_960 977570_960 977580_960 976580_960
479-2162EA 1350 EUR
479-2162 479-2163 479-2164 GREI976561_960 GREI976570_10 GREI976567_960 GREI977561_960 GREI977570_960 GREI977580_960 GREI976580_960
Biobanking tubes, Cryo.s™
Viales
Cryo.s™ biobanking tubes are used for storage of tissue, cells, fungi, bacteria, spores, cellular extracts or body fluids at ultra-low temperature for research and development purposes in a very compact format.
  • Pre-racked
  • Non-capped tubes are non-sterile
  • Conserves upto 30% freezer space
Precaución: Must not be stored in the liquid phase of liquid nitrogen, but only in the gas phase. Not intended for any application in the context of reproductive medicine.
Order Now
ESPECIFICACIONES
Environmentally Preferable Low Manufacturing Impact
Certificación ambiental ISO 14001 (241002); ISO 50001 (241002)