Fmoc-L-4,4'-biphenylalanine 95%

Proveedor: Thermo Fisher Scientific

H52002.MD H52002.03 H52002.06
H52002.MDEA 83.7 EUR
H52002.MD H52002.03 H52002.06
Fmoc-L-4,4'-biphenylalanine 95%
Fmoc-L-4,4'-biphenylalanine

Purity: 95% CAS No.: 199110-64-0 Molecular Formula: C30H25NO4 Formula Weight: 463.52 MDL No.: MFCD00191198

Formula: C₃₀H₂₅NO₄
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00191198
Núm. CAS: 199110-64-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR