Boc-L-β-homoaspartic acid-γ-benzyl ester 95%

Proveedor: Thermo Fisher Scientific

H52191.MD H52191.03
H52191.MDEA 160 EUR
H52191.MD H52191.03
Boc-L-β-homoaspartic acid-γ-benzyl ester 95%
Boc-L-beta-homoaspartic acid-gamma-benzyl ester

Purity: 95% CAS No.: 254101-10-5 Molecular Formula: C17H23NO6 Formula Weight: 337.37 Beilstein Reference: 8430789 MDL No.: MFCD01862861

Formula: C₁₇H₂₃NO₆
Temperatura de almacenaje: Ambiente
Número MDL: MFCD01862861
Núm. CAS: 254101-10-5

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR