N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate ≥98%

Proveedor: Thermo Fisher Scientific

H54402.MB H54402.MC
H54402.MBEA 69.1 EUR
H54402.MB H54402.MC
N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate ≥98%
N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate

Purity: 98+% CAS No.: 146998-31-4 Molecular Formula: C19H18N4O4 Formula Weight: 366.37 Storage Temperature: -30°C to -10°C MDL No.: MFCD00467415

Formula: C₁₉H₁₈N₄O₄
Número MDL: MFCD00467415
Núm. CAS: 146998-31-4

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR