Boc-DAP(Z)-OH·DCHA 98%

Proveedor: Thermo Fisher Scientific

H61558.06
H61558.06EA 155 EUR
H61558.06
Boc-DAP(Z)-OH·DCHA 98%
Boc-DAP(Z)-OH·DCHA

Purity: 98% CAS No.: 65710-58-9 Molecular Formula: C16H22N2O6.C12H23N Formula Weight: 385.54 Physical Form: Solid Beilstein Reference: 4100906 MDL No.: MFCD00236880

Formula: C₂₈H₄₅N₃O₆
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00236880
Núm. CAS: 65710-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR