H-Lys(boc)-OtBu·HCl 95%

Proveedor: Thermo Fisher Scientific

H62993.03 H62993.06
H62993.03EA 129 EUR
H62993.03 H62993.06
H-Lys(boc)-OtBu·HCl 95%
H-Lys(boc)-OtBu·HCl

Purity: 95% CAS No.: 13288-57-8 Molecular Formula: C15H30N2O4.HCl Formula Weight: 338.87 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White MDL No.: MFCD00038899

Formula: C₁₅H₃₁ClN₂O₄
Número MDL: MFCD00038899
Núm. CAS: 13288-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR