(2R,2'R)-Dimethyl-3,3'-disulphanediylbis(2-aminopropanoate) dihydrochloride 98%

Proveedor: Thermo Fisher Scientific
Atención

H63234.03 H63234.06 H63234.14
H63234.03EA 32.9 EUR
H63234.03 H63234.06 H63234.14
(2R,2'R)-Dimethyl-3,3'-disulphanediylbis(2-aminopropanoate) dihydrochloride 98%
(2R,2'R)-Dimethyl-3,3'-disulphanediylbis(2-aminopropanoate) dihydrochloride

Purity: 98% CAS No.: 32854-09-4 Molecular Formula: C8H16N2O4S2.2HCl Formula Weight: 341.28 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 182-183° dec Beilstein Reference: 4727524 Optical Rotation: -36°(c=4 in methanol) MDL No.: MFCD00012490

Formula: C₈H₁₈Cl₂N₂O₄S₂
Pto. de fusión: 182…183 °C (desc.)
Temperatura de almacenaje: Nevera
Número MDL: MFCD00012490
Núm. CAS: 32854-09-4
EINECS: 251-261-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR