Nω-Nitro-L-arginine methyl ester hydrochloride 98%

Proveedor: Thermo Fisher Scientific

Sinónimos: N-ω-Nitro-L-Arginine Methyl Ester Hydrochloride, L-NAME Hydrochloride, L-NAME HCl, H-Arg(NO2)-OMe.HCl, (S)-Methyl 2-amino-5-(3-nitroguanidino)pentanoate hydrochloride

H63666.06 H63666.14
H63666.06EA 51.2 EUR
H63666.06 H63666.14
Nω-Nitro-L-arginine methyl ester hydrochloride 98%
Nomega-Nitro-L-arginine methyl ester hydrochloride

Purity: 98% CAS No.: 51298-62-5 Molecular Formula: C7H15N5O4.HCl Formula Weight: 269.69 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White to off-white Melting Point: 159-161° Beilstein Reference: 3744166 Optical Rotation: +15°(c=3 in methanol) MDL No.: MFCD00039052

Formula: C₇H₁₅N₅O₄
Pto. de fusión: 159…161 °C
Temperatura de almacenaje: Congelador
Número MDL: MFCD00039052
Núm. CAS: 51298-62-5
EINECS: 257-116-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR