Boc-Abu-OH 95%

Proveedor: Thermo Fisher Scientific

Sinónimos: (S)-2-(tert-Butoxycarbonylamino)butyric acid, Boc-L-2-aminobutanoic acid, BOC-L-α-Aminobutyric acid, Boc-L-Abu-OH, (S)-2-(Boc-amino)butyric acid

H63686.03 H63686.06 H63686.14
H63686.03EA 52.5 EUR
H63686.03 H63686.06 H63686.14
Boc-Abu-OH 95%
Boc-Abu-OH

Purity: 95% CAS No.: 34306-42-8 Molecular Formula: C9H17NO4 Formula Weight: 203.24 Physical Form: Solid Beilstein Reference: 6801706 Optical Rotation: -18°(c=1 in methanol) MDL No.: MFCD00037267

Formula: C₉H₁₇NO₄
MW: 203.238 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00037267
Núm. CAS: 34306-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR