H-Aib-OtBu·HCl 98%

Proveedor: Thermo Fisher Scientific

H63817.03 H63817.06
H63817.03EA 181 EUR
H63817.03 H63817.06
H-Aib-OtBu·HCl 98%
H-Aib-OtBu·HCl

Purity: 98% CAS No.: 4512-32-7 Molecular Formula: C8H17NO2.HCl Formula Weight: 195.69 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White MDL No.: MFCD08272284

Formula: C₈H₁₇NO₂
Número MDL: MFCD08272284
Núm. CAS: 4512-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR