(2-Methyl-3-nitrophenyl)acetic acid 96%

Proveedor: Thermo Fisher Scientific
Atención

H64810.06 H64810.14
H64810.06EA 28.3 EUR
H64810.06 H64810.14
(2-Methyl-3-nitrophenyl)acetic acid 96%
(2-Methyl-3-nitrophenyl)acetic acid

Purity: 96% CAS No.: 23876-15-5 Molecular Formula: C9H9NO4 Formula Weight: 195.17 Storage Temperature: 2°C to 8°C Physical Form: Solid Melting Point: 132-136° Beilstein Reference: 2581973 MDL No.: MFCD07782132

Formula: C₉H₉NO₄
Pto. de fusión: 132…136 °C
Número MDL: MFCD07782132
Núm. CAS: 23876-15-5

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR