N-Boc-D-aspartic acid-1-methyl ester 97%

Proveedor: Thermo Fisher Scientific

H66573.14 H66573.06
H66573.14EA 310 EUR
H66573.14 H66573.06
N-Boc-D-aspartic acid-1-methyl ester 97%
N-Boc-D-aspartic acid-1-methyl ester

Purity: 97% CAS No.: 137130-65-5 Molecular Formula: C10H17NO6 Formula Weight: 247.25 Physical Form: Powder Appearance: White

Formula: C₁₀H₁₇NO₆
Temperatura de almacenaje: Ambiente
Número MDL: MFCD03094776
Núm. CAS: 137130-65-5

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR