Powder pillows, Chloride 2 indicator
Proveedor: Hach
104399 105766
HACH104399EA 57.8 EUR
HACH104399 HACH105766
Powder pillows, Chloride 2 indicator
Kits para test medioambiente
Powder reagent in sealed package used as indicator for the argentometric titration of chloride.
Precaución: Note: Product contains potsassium chromate. Please read MSDS prior to usage.