Reagent powder pillows for determination of molybdenum, MolyVer®
Proveedor: Hach
1414669 1414869 1417869
HACH1414669EA 80.8 EUR
HACH1414669 HACH1414869 HACH1417869
Reagent powder pillows for determination of molybdenum, MolyVer®
Kits para test medioambiente
For determination of high range molybdenum by the mercaptoacetic acid method.