Comparator kits for water testing
Proveedor: Hach
1416100 146700 2064400 1419301 145401 2518050
705-0055EA 183 EUR
705-0055 HACH146700 HACH2064400 HACH1419301 705-0068 HACH2518050
Comparator kits for water testing
Kits para test medioambiente
Qualitative results using traditional colour comparator disc methods.
  • Portable - ideal for field testing
  • Simple fast method
  • Kits contain all required reagents and apparatus in a rugged carrying case
Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.