Stoppers, hollow, LDPE
Proveedor: Hach
1448001 1448000
HACH1448001EA 21.7 EUR
HACH1448001 HACH1448000
Stoppers, hollow, LDPE
Tapones Tapones de PE/LDPE/HDPE
Ideally can be used as a small beaker, sample cup for pH meters, flame photometers and automatic analysers.
  • LDPE
  • Interchangeable with rubber stoppers
  • Safer and economical
  • Fits snugly