Sulphide reagents 1 and 2
Proveedor: Hach
181632 181732 181749
HACH181632EA 35.1 EUR
HACH181632 HACH181732 HACH181749
Sulphide reagents 1 and 2
Kits para test medioambiente
Used for sulphide determination by the methylene blue method.
  • Range upto 800 µg/L
  • Used with visual test kits