Erlenmeyer flasks, polymethylpentene
Proveedor: Hach
2089846
HACH2089846EA 33.7 EUR
HACH2089846
Erlenmeyer flasks, polymethylpentene
Matraces Matraces Erlenmeyer
Ideal for titrations, preparation and storage of media.
  • Chemical resistance Strength
  • Autoclavable
Polymethylpentene flask with polypropylene screwcap features clarity (Do not engage threads during cycle).