Reagent set, glycol test kit
Proveedor: Hach
2446900
HACH2446900EA 679 EUR
HACH2446900
Reagent set, glycol test kit
Kits para test medioambiente
Used for determination of glycol by the EG-1 glycol test kit.
Información de suministro: Reagent set includes: glycol test reagent 1, glycol test reagent 2, glycol test reagent 3 and ethylene glycol - 0.05% solution.