Reducing solution for ORP electrodes
Proveedor: Hach
2965349
HACH2965349EA 47.9 EUR
HACH2965349
Reducing solution for ORP electrodes
Electrolitos Soluciones de almacenamiento para electrodos
Used to condition ORP electrode surfaces for use in reducing sample matrices.