Reference electrodes, Radiometer Analytical
Proveedor: Hach
E21M009 E21M001 E21M002 B20B300
HACHE21M009EA 330 EUR
HACHE21M009 HACHE21M001 HACHE21M002 HACHB20B300
Reference electrodes, Radiometer Analytical
Electrodos Electrodos selectivos de iones (ISE)
Radiometer Analytical reference electrodes are available with a variety of reference systems and liquid junctions.
  • Liquid junction: porous pin
  • Red-Rod technology provides a reproducible, fast and accurate response
  • Calomel reference electrode can be used for special purposes
* These electrodes contain mercury, please dispose according to regulations.
** Screw cap electrodes require an external connection cable for connection to a meter.