AERO CHROME* Crew Performance Surgical Gowns
Proveedor: Halyard
48392 48393 48394 48399 48400 48401
HALY48392EA 240 EUR
HALY48392 HALY48393 HALY48394 HALY48399 HALY48400 HALY48401
AERO CHROME* Crew Performance Surgical Gowns
Prendas desechables
AERO CHROME* CREW Breathable surgical gowns combine unprecedented softness and breathability with maximum protection against fluids and pathogens.
  • Meets AAMI Level 4 standard, providing the highest level of both fluid and microbial protection in the critical zones
  • Proprietary fabric technology allows moisture vapor to escape while maintaining a high performance barrier
  • Designed to keep your OR staff both cool and protected during long, fluid-intensive procedures
  • Highest rating for resistance to ignition from surgical lasers
  • Excellent abrasion and lint