1000 and 1700 series Gastight® syringes for PSD/4 syringe pumps
Proveedor: HAMILTON BONADUZ
5495-50 5495-10 5495-15 5495-20 5495-25 5495-30 5495-35 5495-40 5495-45 5495-55 5495-22 5495-38
HAMI5495-50EA 203 EUR
HAMI5495-50 HAMI5495-10 HAMI5495-15 HAMI5495-20 HAMI5495-25 HAMI5495-30 HAMI5495-35 HAMI5495-40 HAMI5495-45 HAMI5495-55 HAMI5495-22 HAMI5495-38
1000 and 1700 series Gastight® syringes for PSD/4 syringe pumps
Jeringas Jeringas para cromatografía
Syringes for the PSD/4 syringe pump are available with a standard PTFE tipped plunger and with a long life UHMWPE (Ultra High Molecular Weight Poly Ethylene) plunger tip. The stroke length of a PSD/4 syringe is 30 mm.
Syringe terminations:
TLL, PTFE® Luer Lock
This termination has a PTFE, male Luer taper with nickel plated brass locking hub for use with Kel-F needles, metal hub needles and universal connectors. Also, the TLL is used with Hamilton Diluters/Dispensers, OEM applications and manual operations. Autoclavable when disassembled, except on 25 ml and greater syringes. RepeatedMore Product Information