Jeringas Gastight® Serie 1800
Proveedor: HAMILTON BONADUZ
84875 84877 84878 84880 84881 84883 84884 84886 84887 84889 7663-01 7659-01 7662-01 7660-01 7661-01 CAL84887 CAL84877 CAL84875 CAL84881 CAL7659-01 CAL7660-01 CAL7661-01 CAL7662-01
549-1214EA 163 EUR
549-1214 549-1215 549-1216 549-1217 549-1218 549-1219 549-1220 549-1221 549-1222 549-1223 HAMI7663-01 HAMI7659-01 HAMI7662-01 HAMI7660-01 HAMI7661-01 HAMICAL84887 HAMICAL84877 HAMICAL84875 HAMICAL84881 HAMICAL7659-01 HAMICAL7660-01 HAMICAL7661-01 HAMICAL7662-01
Jeringas Gastight® Serie 1800
Jeringas Jeringas para cromatografía
The 1800 series is the reinforced plunger version of the 1700 series syringe. Attached to the flange of the syringe is a removable blue syringe holder that prevents heat transfer, dispense inaccuracies and plunger blow outs. It also provides support to small volume plungers that are prone to bending during injection.
  • Reinforced, protected plunger
  • Ideal for dispensing both liquids and gases in volumes from 1 up to 500 µl
  • Gas sterilisable
  • Stainless steel, borosilicate glass and PTFE fluid path
They have a precision machined PTFE plunger tip whichMore Product Information