Syringes, SampleLock™
Proveedor: HAMILTON BONADUZ
81656 81056 86326 86336 86346 81256 81356 81456 81556 80956 81156 CAL81156
HAMI81656EA 383 EUR
HAMI81656 HAMI81056 HAMI86326 HAMI86336 HAMI86346 HAMI81256 HAMI81356 HAMI81456 HAMI81556 HAMI80956 HAMI81156 HAMICAL81156
Syringes, SampleLock™
Jeringas Jeringas para cromatografía
These syringes allow the collection, storage, transport and analysis of liquid or gaseous samples without the danger of evaporative loss or environmental contamination.
  • Not autoclavable, gas sterilisable
  • An easy-to-use twist valve
  • A positive rear stop prevents loss of sample and plunger blowout
  • Fluid path includes SST, borosilicate glass and PTFE
The sample lock termination incorporates an on/off syringe valve with removable needle. This termination is used for headspace, environmental sample collection and storage, pre-pressurization of gaseousMore Product Information