Vibration spatula
Proveedor: HAMMACHER KARL
HSN462-22
HAMMHSN462-22EA 57.9 EUR
HAMMHSN462-22
Vibration spatula
Espátulas
Vibration spatula with acid-proof plastic handle, PTFE-coated. For fine dosing.