Weighing papers
Proveedor: HEATHROW
HS120116
HEATHS120116EA 59.8 EUR
HEATHS120116
Weighing papers
Papeles de pesaje
These weighing papers are nitrogen-free for a very smooth surface and good sample recovery.
  • Disposable
  • Strong enough to support dense material
Colour: White