Microtube racks, 96-well
Proveedor: HEATHROW
HS120243
HEATHS120243EA 95.5 EUR
HEATHS120243
Microtube racks, 96-well
Gradillas Tube Racks
These microtube racks are made from reinforced nylon.
  • Superior chemical- and heat-resistance
  • Handles provide easy and safe transportation
  • Stackable two formats available for smaller and larger requirements
  • Have 11,1 mm Ø wells
  • Autoclavable