Cell counters
Proveedor: HEATHROW
HS5360A HS5360B
HEATHS5360AEA 461 EUR
HEATHS5360A HEATHS5360B
Cell counters
Contadores de células Contadores manuales
These laboratory cell counters can measure cells such as basophils, eosinophils, myelocytes, juvenile, etc.
  • Each key records up to 999
  • Alarm at one hundred totals
  • Red and white buttons with grey housing
They can measure the number of cells such as Basophils (BASO), Eosinophils (EOSIN), Myelocytes (MYELO), Juvenile (JUVEN), Stab., Seg., Lymph., Mono., and total.