Blood sedimentation apparatus
Proveedor: GLASWARENFABRIK KARL HECHT
40367010
HECH40367010EA 210 EUR
HECH40367010
Blood sedimentation apparatus
Sistemas para diagnóstico Clinical Instruments
Blood sedimentation apparatus stand of stainless steel with tilting device for rapid sedimentation.
  • Pipettes with fused graduation 'ASSISTENT-PERMANENT'
Información para pedidos: Complete with blood sedimentation pipettes, blood mixing glasses with rubber stopper, special rubber caps (macro method only) and directions for use.
Envase: In a box.