Dropping bottles TK, glass
Proveedor: GLASWARENFABRIK KARL HECHT
41319012 41318012 41318006 41319006
HECH41319012EA 11.4 EUR
HECH41319012 HECH41318012 HECH41318006 HECH41319006
Dropping bottles TK, glass
Frascos Frascos de goteo
Glass.
  • Good chemical and physical properties
  • For use in applications that do not involve high thermal stresses
  • Amber coloured to protect light sensitive substances
Dropping bottles avalaible in clear and amber format with flat glass stopper.