Rubber Stoppers
Proveedor: GLASWARENFABRIK KARL HECHT
41590002 41590011 41590014 41590017 41590020 41590023 41590032 41590035 41590038 41590041 41590044 41590047 41590053 41590056 41590059 92159111490
HECH41590002EA 0.7 EUR
HECH41590002 HECH41590011 HECH41590014 HECH41590017 HECH41590020 HECH41590023 HECH41590032 HECH41590035 HECH41590038 HECH41590041 HECH41590044 HECH41590047 HECH41590053 HECH41590056 HECH41590059 HECH92159111490
Rubber Stoppers
Tapones Tapones de Goma
DIN 12871, made of natural rubber, hardness 40 Shore A.