Measuring cylinders, tall form, class B, borosilicate glass
Proveedor: GLASWARENFABRIK KARL HECHT
42180030 42180024 42180060 42180066
HECH42180030EA 11.7 EUR
HECH42180030 HECH42180024 HECH42180060 HECH42180066
Measuring cylinders, tall form, class B, borosilicate glass
Probetas Probetas graduadas
Borosilicate glass, tall form, class B.
  • Calibrated to contain 'ln'
  • Hexagonal glass base, with spout
  • Fused brown graduations
Certificaciones: DIN EN ISO 4788