Graduated pipettes, with piston
Proveedor: GLASWARENFABRIK KARL HECHT
42548020 42548048 42548052 42548036 42548056 42548044
HECH42548020EA 24.5 EUR
HECH42548020 HECH42548048 HECH42548052 HECH42548036 HECH42548056 HECH42548044
Graduated pipettes, with piston
Pipetas Pipetas graduadas
Safety measuring pipettes with interchangeable ground-in piston, standard type.
  • Tolerance 1,5 times higher than the official tolerance of class AS
  • Fused amber graduation 'ASSISTENT-PERMANENT'
  • Zero at top, calibrated
  • Fire-polished tip
Envase: Each pipette is packed in a plastic tube.