Electronic max/min thermometer
Proveedor: GLASWARENFABRIK KARL HECHT
43656010
HECH43656010EA 21.5 EUR
HECH43656010
Electronic max/min thermometer
Termómetros Termómetros digitales
Maximum-minimum electronic thermometer used for measuring indoor and outdoor temperature.
  • Present temperature/highest and lowest day temperature
  • For hanging or standing
  • Extra thin cable for easily leading through window seals
  • Waterproof sensor for outdoor measurements
* Indoor
** Outdoor
Información de suministro: Incl. 1,5 V AA battery.