Hei-PLATE Mix 'n' Heat Core Magnetic Stirrers
Proveedor: Heidolph Instruments GmbH & Co.KG
506-11100-00 506-12100-00
HEID506-11100-00EA 948 EUR
HEID506-11100-00 HEID506-12100-00
Hei-PLATE Mix 'n' Heat Core Magnetic Stirrers
Imánes de agitación Agitadores magnéticos con placas calefactoras
In accordance with the motto 'simply stir and heat', our entry-level model Hei-PLATE Mix 'n' Heat Core was designed with two rotary knobs to allow direct access to the two main functions. The speed and temperature are set using a scale. The maximum values are 1400 rpm and 300 °C.
  • Patented Kera-Disk® footprint made of anodised aluminum
  • Chemical-resistant
  • Scratch-proof
  • Has a diameter of 145 mm
This magnetic stirrer is very easy to use and is equipped with the most important safety features. The large safety distance between the heating plate and theMore Product Information