Tubing, silicone, HelixMark®
Proveedor: Freudenberg Medical Europe GmbH
60-795-22 60-795-36 60-795-05 60-795-30 60-795-21
228-0849EA 150 EUR
228-0849 228-0850 HELI60-795-05 HELI60-795-30 HELI60-795-21
Tubing, silicone, HelixMark®
Tuberías Tubo de transferencia
Silicone, translucent, platinum-cured.
  • Can be sterilised by autoclaving or ethylene oxide as well as by gamma radiation within the user's validation cycles and exposure levels
  • Manufactured according to GMP in an FDA-registered production facility for medical products
  • Compliant with the biocompatibility requirements of USP, Class VI
Biomedical/pharmaceutical grade tubing, manufactured by Nusil platinum-cured material. The special manufacturing process minimises flow degradation over extended periods.
Hardness (Shore durometer A): 50
Density: 1.14