Medifuge™ Small Centrifuge, Bench
Proveedor: Thermo Scientific
75008800 75008801
521-2225EA 1560 EUR
521-2225 HERA75008801
Medifuge™ Small Centrifuge, Bench
Centrífugas Centrífugas de sobremesa
This compact centrifuge is supplied with a unique DualSpin™ rotor featuring a 2-in-1 hybrid design. This feature enables users to easily change between fixed angle and swinging buckets using the same rotor.
  • Lightweight composite rotor is easy to handle and saves energy during acceleration/deceleration
  • Rotor can accommodate 1,4 to 15 ml tubes with two spacer options
  • Large, brightly-lit display is easy to read and fast one-click lid closure simplifies operation
  • Four customisable programs to quickly run routine protocols
  • Three selectable deceleration profiles