Accessories for ultra-low temperature freezers
Proveedor: Thermo Scientific
FLN300D FCO300D FLN4567 FLN300V FLN300A FCO4567 FCO300W FLN300W LN4567 FCO300A FCO300V FCO456LEV FLN456LEV LNTSXU
HERAFLN300DEA 2540 EUR
HERAFLN300D HERAFCO300D HERAFLN4567 471-1234 HERAFLN300A HERAFCO4567 HERAFCO300W HERAFLN300W HERALN4567 HERAFCO300A 471-1235 471-1232 471-1233 HERALNTSXU
Accessories for ultra-low temperature freezers
Congeladores Ultracongeladores
Ensure optimal security for critical samples by using Thermo Scientific™ CO₂ and LN2 Backup Systems for Ultra-Low Temperature Freezers, for use with Thermo Scientific™ TSX, Forma™, HERAfreeze™, Revco™ and TS-Series freezers.
  • LN₂ backup system maintains temperature down to –80 °C with liquid nitrogen, CO₂ backup system maintains temperature down to –67 °C with CO₂
LN₂ backup system: With LN₂ emergency cooling, a mobile tank is sufficient if it is ensured that the fill level is regularly replenished. The supply pressure must be less than ~2,5 bar, thisMore Product Information