Capillary columns, DB-1701
Proveedor: Agilent
121-0713 122-0712 122-0713 122-0731 122-0732 122-0733 122-0761 122-0762 122-0763 122-0766 123-0712 123-0713 123-0731 123-0732 123-0733 123-0753 123-0762 123-0763 123-0763E 125-0712 125-0731 125-0732 125-0732E 125-0733 125-0737 127-0722 127-0723 122-0732E 123-0732E 123-0733E 125-0732G 125-0762 123-0753E 123-0762E 125-0712E 125-0762E 127-0723E 122-0733E 122-0763E 121-0722 121-0722LTM 122-0713LTM 122-0732LTM 122-0733LTM 123-0712LTM 125-0712LTM
HEWL121-0713EA 393 EUR
HEWL121-0713 HEWL122-0712 HEWL122-0713 HEWL122-0731 HEWL122-0732 HEWL122-0733 HEWL122-0761 HEWL122-0762 HEWL122-0763 HEWL122-0766 HEWL123-0712 HEWL123-0713 HEWL123-0731 HEWL123-0732 HEWL123-0733 HEWL123-0753 HEWL123-0762 HEWL123-0763 HEWL123-0763E HEWL125-0712 HEWL125-0731 HEWL125-0732 HEWL125-0732E HEWL125-0733 HEWL125-0737 HEWL127-0722 HEWL127-0723 HEWL122-0732E HEWL123-0732E HEWL123-0733E HEWL125-0732G HEWL125-0762 HEWL123-0753E HEWL123-0762E HEWL125-0712E HEWL125-0762E HEWL127-0723E HEWL122-0733E HEWL122-0763E HEWL121-0722 HEWL121-0722LTM HEWL122-0713LTM HEWL122-0732LTM HEWL122-0733LTM HEWL123-0712LTM HEWL125-0712LTM
Capillary columns, DB-1701
Columnas de cromatografía
These columns contain (14%-Cyanopropyl-phenyl)-methylpolysiloxane.
  • Low/mid-polarity
  • Bonded and cross-linked
  • Solvent rinsable