Capillary columns, DB-1
Proveedor: Agilent
122-1062 122-106E 123-105C 121-1012 121-1013 121-101A 121-1022 121-1023 121-1043 122-1011 122-1012 122-1013 122-1022 122-1031 122-1032 122-1032E 122-1033 122-103E 122-1052 122-1061 122-1063 122-10AE 123-1011 123-1012 123-1013 123-1014 123-1015 123-1022 123-1026 123-1027 123-102F 123-1031 123-1032 123-1033 123-1033E 123-1034 123-1035 123-103E 123-1052 123-1055 123-1056 123-105F 123-1061 123-1062 123-1063 123-1065 123-106B 123-106E 123-106G 125-1032 125-1035 126-1012 126-1013 127-100A 127-1012 127-1012E 127-1013 127-1013E 127-1022 127-1023 127-1043 127-1046 128-1012 128-1022 128-1034 128-1052 122-1033E 122-10G3 123-103B 123-1064E 123-1065E 127-1022E 12A-1015 121-1012E 123-1062E 123-1063E 123-106BE 127-1023E 121-1022E 121-1043E 123-1032LTM 121-1012LTM 121-1013LTM 121-101ALTM 121-1022LTM 121-1023LTM 122-1012LTM 122-1022LTM 122-1032LTM 122-1033LTM 122-103ELTM 123-1011LTM 123-1012LTM 123-1015LTM 123-1033LTM 123-1035LTM 123-103BLTM 123-103ELTM 127-100ALTM 127-1013LTM 127-1023LTM 127-1046E 128-1012LTM 128-1022LTM 12A-1015LTM
HEWL122-1062EA 1292 EUR
HEWL122-1062 HEWL122-106E HEWL123-105C HEWL121-1012 HEWL121-1013 HEWL121-101A HEWL121-1022 HEWL121-1023 HEWL121-1043 HEWL122-1011 558-0360 HEWL122-1013 HEWL122-1022 HEWL122-1031 558-0361 HEWL122-1032E HEWL122-1033 HEWL122-103E HEWL122-1052 HEWL122-1061 558-0362 HEWL122-10AE HEWL123-1011 HEWL123-1012 HEWL123-1013 HEWL123-1014 HEWL123-1015 HEWL123-1022 HEWL123-1026 HEWL123-1027 HEWL123-102F HEWL123-1031 HEWL123-1032 HEWL123-1033 HEWL123-1033E HEWL123-1034 HEWL123-1035 HEWL123-103E HEWL123-1052 HEWL123-1055 HEWL123-1056 HEWL123-105F HEWL123-1061 HEWL123-1062 HEWL123-1063 HEWL123-1065 HEWL123-106B HEWL123-106E HEWL123-106G HEWL125-1032 558-0371 HEWL126-1012 HEWL126-1013 HEWL127-100A HEWL127-1012 HEWL127-1012E HEWL127-1013 HEWL127-1013E HEWL127-1022 HEWL127-1023 HEWL127-1043 HEWL127-1046 HEWL128-1012 HEWL128-1022 HEWL128-1034 HEWL128-1052 HEWL122-1033E HEWL122-10G3 HEWL123-103B HEWL123-1064E HEWL123-1065E HEWL127-1022E HEWL12A-1015 HEWL121-1012E HEWL123-1062E HEWL123-1063E HEWL123-106BE HEWL127-1023E HEWL121-1022E HEWL121-1043E HEWL123-1032LTM HEWL121-1012LTM HEWL121-1013LTM HEWL121-101ALTM HEWL121-1022LTM HEWL121-1023LTM HEWL122-1012LTM HEWL122-1022LTM HEWL122-1032LTM HEWL122-1033LTM HEWL122-103ELTM HEWL123-1011LTM HEWL123-1012LTM HEWL123-1015LTM HEWL123-1033LTM HEWL123-1035LTM HEWL123-103BLTM HEWL123-103ELTM HEWL127-100ALTM HEWL127-1013LTM HEWL127-1023LTM HEWL127-1046E HEWL128-1012LTM HEWL128-1022LTM HEWL12A-1015LTM
Capillary columns, DB-1
Columnas de cromatografía
These columns contain 100% dimethylpolysiloxane.
  • Non-polar
  • Very low bleed characteristics, ideal for GC/MS
Max. temperature: -60 to 325/350 °C