Capillary columns, Biodiesel
Proveedor: Agilent
123-BD34
HEWL123-BD34EA 822 EUR
HEWL123-BD34
Capillary columns, Biodiesel
Columnas de cromatografía
Biodiesel capillary GC columns are designed and application optimised for the analysis of biodiesel to meet ASTM and CEN testing standards. These columns are used for the measurement of residual methanol in B100.
  • Bonded and cross-linked
  • Solvent rinsable
  • Temperature limits from 20 to 260/280 °C