Capillary columns, DB-23
Proveedor: Agilent
121-2323 122-2312 122-2331 122-2332 122-2361 122-2362 123-2332 123-2362 125-2312 125-2332 123-2332E 122-2332E 122-2361E 122-2362E 122-2332LTM
HEWL121-2323EA 630 EUR
HEWL121-2323 HEWL122-2312 HEWL122-2331 HEWL122-2332 HEWL122-2361 HEWL122-2362 HEWL123-2332 HEWL123-2362 HEWL125-2312 HEWL125-2332 HEWL123-2332E HEWL122-2332E HEWL122-2361E HEWL122-2362E HEWL122-2332LTM
Capillary columns, DB-23
Columnas de cromatografía
These columns are designed for separation of fatty acid methyl esters (FAMEs).
  • Solvent rinsable
  • Excellent resolution for cis- and trans-isomers
  • Bonded and cross-linked
(50%-Cyanopropyl)-methylpolysiloxane, highly polar
Max. temperature: 250/260 °C
Suitable replacement for HP-23.