Spectrophotometer cuvettes
Proveedor: Agilent
5061-3391 5063-6558 5063-6559 5063-6560 5063-6561
HEWL5061-3391EA 634 EUR
HEWL5061-3391 HEWL5063-6558 HEWL5063-6559 HEWL5063-6560 HEWL5063-6561
Spectrophotometer cuvettes
Cubetas Cubetas para espectrofotómetros
Spectrophotometer cuvettes with beam height of 15 mm.