Agua, grado ultrapuro for CE

Proveedor: Agilent

Sinónimos: Hydrogen oxide

5062-8578
HEWL5062-8578EA 110 EUR
HEWL5062-8578
Agua, grado ultrapuro for CE
Agua

Premade buffers help eliminate the time-consuming buffer preparation process. All Agilent buffers and reagents are designed to meet the stringent demands of CE.;To maintain high reproducibility, all buffer solutions are standardized to 0.02 pH units under "class 10" clean room conditions. Superior quality control ensures reproducible results bottle-to-bottle and batch-to-batch.


  • Manufactured under GLP/GMP conditions in ISO9001 facilities
  • Chemicals are all electrophoresis grade, with nearly all ionic and organic impurities removed
  • Solutions are prefiltered through 0.2 mm filters to ensure removal of particulates
  • All buffer solutions are standardized to 0.02 pH units in accredited laboratories
  • Each buffer shipped with assay information and verification of purity

Formula: H₂O
Pto. Ebullición: 100 °C (1013 hPa)
Pto. de fusión: 0 °C
Densidad: 1 g/cm³ (4 °C)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00011332
Núm. CAS: 7732-18-5
EINECS: 231-791-2
Índice Merck: 12,10175

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR