1290 valve head capillaries
Proveedor: Agilent
5067-5749 5067-5750 5067-5109 5067-4684 5067-4686 5067-4687 5067-4689 5067-5110 5067-5106 5067-5111 5067-5112 5067-5709 5067-5713 5067-5113 5500-1199 5500-1201 5500-1202 5500-1203 5500-1204 5500-1242 5500-1243 5500-1333 5500-1334 5500-1303 5500-1302 5500-1301 5500-1300 5500-1335 5500-1336 5500-1246
HEWL5067-5749EA 54 EUR
HEWL5067-5749 HEWL5067-5750 HEWL5067-5109 HEWL5067-4684 HEWL5067-4686 HEWL5067-4687 HEWL5067-4689 HEWL5067-5110 HEWL5067-5106 HEWL5067-5111 HEWL5067-5112 HEWL5067-5709 HEWL5067-5713 HEWL5067-5113 HEWL5500-1199 HEWL5500-1201 HEWL5500-1202 HEWL5500-1203 HEWL5500-1204 HEWL5500-1242 HEWL5500-1243 HEWL5500-1333 HEWL5500-1334 HEWL5500-1303 HEWL5500-1302 HEWL5500-1301 HEWL5500-1300 HEWL5500-1335 HEWL5500-1336 HEWL5500-1246
1290 valve head capillaries
Tuberías Chromatography Tubing
A comprehensive range of capillaries and fittings manufactured to Agilent's highest level of quality to protect the integrity of the chromatographic results.
  • Best fit to each 1290 valve head for optimum chromatographic performance
  • Flexible stainless steel capillaries for increased usability without scarifying durability
  • Ultra high pressure tightness
  • Leak free connection